
Dehydroacetic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:520-45-6
:771-03-9
Formula:C₈H₈O₄
MDL Number:MFCD00066709
:168.15 g/mol
Merck Index:13,02885
Boiling Pt:333 °C (1013 hPa)
Melting Pt:109…112 °C
Flash Pt:157 °C
EINECS:208-293-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Cream to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:109 to 112 °C
- Sulfated ash:≤0.1 % (700 °C, 2 h)
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together