Iron(II) sulphide, fused sticks
Catalog # ACRO233300010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Appearance:Grey-black Sticks
- Assay:(S) about 25%
Specification Test Results
About this item
:
:87.91 g/mol
Merck Index:12,04106
Melting Pt:1195 °C
Density:4.84 g/cm³
EINECS:215-268-6
ADR:9,III
CAS Number:1317-37-9
Formula:FeS
MDL Number:MFCD00011013