Tungsten disulphide, Sigma-Aldrich®
Catalog # SIAL243639-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
About this item
Synonyms: Tungsten sulphide, Tungsten (IV) sulphide
:
:247.97 g/mol
EINECS:235-243-3
CAS Number:12138-09-9
Formula:S₂W
MDL Number:MFCD00011467
Density:7.5 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100