Tungsten(VI) fluoride, Sigma-Aldrich®
Catalog # SIAL399108-225G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:225 g
- Seq:3525
Specifications
About this item
:
:297.83 g/mol
Merck Index:12,09946
Boiling Pt:17.5 °C (1013 hPa)
Melting Pt:3 °C
Density:12.9 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:232-029-1
CAS Number:7783-82-6
Formula:F₆W