
Chromium(III) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10025-73-7
:
Formula:Cl₃Cr
MDL Number:MFCD00010948
:158.35 g/mol
Merck Index:12,02278
Melting Pt:1152 °C
Density:2.76 g/cm³ (20 °C)
EINECS:233-038-3
ADR:8,III
:M-Clarity™ quality level = MQ100
UN:3260
:GHS07
Storage Temperature:Ambient
Frequently Bought Together