Vanadium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10580-52-6
:
Formula:Cl₂V
:121.85 g/mol
Boiling Pt:1506 °C (1013 hPa)
Melting Pt:1027 °C
Density:3.23 g/cm³ (20 °C)
EINECS:234-176-7
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ100
Frequently Bought Together