Sodium bitartrate monohydrate ≥99.0% (by titrimetric analysis), BioUltra, Sigma-Aldrich®
Catalog # SIAL71679-250G
Supplier: Merck
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:≥99.0% (T)
- chloride (Cl⁻):≤50 mg/kg
- sulphate (SO₄)²⁻:≤50 mg/kg
- Al:≤5 mg/kg
- As:≤0.1 mg/kg
- Ba:≤5 mg/kg
- Bi:≤5 mg/kg
- Ca:≤50 mg/kg
- Cd:≤5 mg/kg
- Co:≤5 mg/kg
- Cr:≤5 mg/kg
- Cu:≤5 mg/kg
- Fe:≤5 mg/kg
- K:≤50 mg/kg
- Li:≤5 mg/kg
- MG:≤5 mg/kg
- Mn:≤5 mg/kg
- Mo:≤5 mg/kg
- Ni:≤5 mg/kg
- PB:≤5 mg/kg
- Sr:≤5 mg/kg
- Zn:≤5 mg/kg
Specification Test Results
About this item
:
:190.083 g/mol
EINECS:208-400-9
CAS Number:6131-98-2
Formula:C₄H₅NaO₆·H₂O
MDL Number:MFCD00150038