

Zirconium tetrachloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10026-11-6
:
Formula:Cl₄Zr
MDL Number:MFCD00011306
:233.03 g/mol
Merck Index:12,10307
Boiling Pt:3577 °C (1013 hPa)
Melting Pt:437 °C
Density:2.8 g/cm³ (20 °C)
UN:2503
ADR:8,III
:M-Clarity™ quality level = MQ100
EINECS:233-058-2
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together