Order Entry
United Kingdom
ContactUsLinkComponent
 
Scandium(III) chloride, Sigma-Aldrich®
Catalog # SIAL686077-1G
Supplier:  Merck
CAS Number:  
Scandium(III) chloride, Sigma-Aldrich®
Catalog # SIAL686077-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.31 g/mol
Melting Pt:940 °C
Density:1.67 g/cm³ (940 °C)
 :M-Clarity™ quality level = MQ100
EINECS:233-799-1
CAS Number:10361-84-9
Formula:Cl₃Sc
MDL Number:MFCD00011221