

Cadmium selenide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1306-24-7
:
Formula:CdSe
MDL Number:MFCD00010917
:191.37 g/mol
Merck Index:12,01663
Melting Pt:1268 °C
Density:5.81 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-148-3
ADR:6.1,II
:M-Clarity™ quality level = MQ100
UN:3283
:GHS07
:GHS09
Frequently Bought Together