Zinc chloride 98+%, extra pure
:7646-85-7
:
:ZnCl₂
:MFCD00011295
:136.3 g/mol
:13,10185
:732 °C (760 mmHg)
:293 °C
:2,91 g/cm³ (20 °C)
:Ambient
:231-592-0
:2331
:8,III
:GHS07
:GHS05
:GHS09
- Appearance:White to off-white Crystalline powder or crystals and/or chunks
- Titration Complexometric:≥98.0 %
- Cadmium (Cd):≤10 ppm
- Copper (Cu):≤10 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤10 ppm
- Manganese (Mn):≤30 ppm
Frequently Bought Together