Mercury(II) bromide 99+%
: Mercuric bromide, Mercury dibromide
- :7789-47-1
- :Br₂Hg
- :MFCD00011040
- :360.4 g/mol
- :13,05900
- :320 °C (1013 hPa)
- :236 °C
- :6,1 g/cm³ (20 °C)
- :Ambient
- :232-169-3
- :1634
- :6.1,II
: Mercuric bromide, Mercury dibromide
- :7789-47-1
- :Br₂Hg
- :MFCD00011040
- :360.4 g/mol
- :13,05900
- :320 °C (1013 hPa)
- :236 °C
- :6,1 g/cm³ (20 °C)
- :Ambient
- :232-169-3
- :1634
- :6.1,II
- Appearance:White Crystalline powder
- Assay:≥99.0 %
- Chloride (Cl):≤2000 ppm
- Iron (Fe):≤50 ppm
- Insoluble matter:≤0.05 % (in methanol)
Product Family Options
- Pack typePk
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



