Copper(II) sulphate pentahydrate ≥99%, blue, crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7758-99-8
:
Formula:CuSO₄·5H₂O
MDL Number:MFCD00149681
:249.69 g/mol
Merck Index:12,02722
Melting Pt:200 °C
Density:3.603 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3077
ADR:9,III
:GHS07
:GHS09
Frequently Bought Together