Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Silver(I) oxide

Silver(I) oxide

Oxidizing
Corrosive
Marketsource
Supplier:   Roth Carl
Silver(I) oxide
Supplier:  Roth Carl
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  di-Silver oxide, Disilver oxide, Silver oxide
  • CAS Number:20667-12-3
  • Formula:Ag₂O
  • MDL Number:MFCD00003404
  • Molecular Weight:231.74 g/mol
  • Merck Index:12,08665
  • Melting Pt:800…830 °C
  • Density:7.14 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:243-957-1
  • UN:1479
  • ADR:5.1,II
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource
  • 50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g

    Specifications:
    Marketsource
    Marketsource