Silver(I) oxide
50 g
Catalog # ABCRAB101522.0050
Marketsource
Supplier: abcr
Jump to:
Product Details & Documents
Synonyms: di-Silver oxide, Disilver oxide, Silver oxide
- CAS Number:20667-12-3
- Formula:Ag₂O
- MDL Number:MFCD00003404
- Molecular Weight:231.74 g/mol
- Merck Index:12,08665
- Melting Pt:800…830 °C
- Density:7.14 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:243-957-1
- UN:1479
- ADR:5.1,II
Specifications
Specifications
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



