Silver nitrite 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7783-99-5
:
Formula:AgNO₂
MDL Number:MFCD00003413
Merck Index:12,08663
Melting Pt:140 °C
Density:4.45 g/cm³
Storage Temperature:Ambient
EINECS:232-041-7
UN:2627
ADR:5.1,II
:153.87 g/mol
:GHS03
:GHS07
:GHS09
Specification Test Results
- Appearance:Yellow to yellow-green Powder or crystals and/or chunks
- Titration Argentometric:≥98.5 %
Specification Test Results
Frequently Bought Together