Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Acenaphthene, extra pure

Acenaphthene, extra pure

Glass ampoule, 100 mg
Catalog # 122852X
Environment
Supplier:   VWR Chemicals
Acenaphthene, extra pure
Catalog # 122852X
Supplier:  VWR Chemicals
CAS Number:  83-32-9
Acenaphthene, extra pure
Acenaphthene, extra pure

Glass ampoule, 100 mg

CAS Number:  83-32-9
Price
£42.40
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  1,2-Dihydroacenaphthylene
  • CAS Number:83-32-9
  • Formula:C₁₂H₁₀
  • MDL Number:MFCD00003807
  • Molecular Weight:154.21 g/mol
  • Merck Index:13,00029
  • Boiling Pt:279 °C (1013 hPa)
  • Melting Pt:90…93 °C
  • Density:1.024 g/cm³ (20 °C)
  • Flash Pt:135 °C (open cup)
  • Storage Temperature:Ambient
  • EINECS:201-469-6
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.