Praseodymium(III,IV) oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12037-29-5
:
Formula:O₁₁Pr₆
MDL Number:MFCD00011178
:1021.44 g/mol
Melting Pt:2012…2072 °C
:M-Clarity™ quality level = MQ100
EINECS:234-857-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together