Order Entry
United Kingdom
ContactUsLinkComponent
 
Holmium(III) oxide, Sigma-Aldrich®
 :  Merck
 
Holmium(III) oxide, Sigma-Aldrich®
 :  Merck

 

  • Molar Mass:
    377.86 g/mol
  • Hill Formula:
    Ho2O3
  • Chemical Formula:
    Ho2O3

 

Frequently Bought Together