Order Entry
United Kingdom
ContactUsLinkComponent
 
 
1-Methylindole 98+%
 
undefined
 
1-Methylindole 98+%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6050 to 1.6080 (20°C, 589 nm)
  • GC:
    ≥98.0 %

 

 

Frequently Bought Together