5-Aminoindole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5192-03-0
:
Formula:C₈H₈N₂
MDL Number:MFCD00005679
:132.16 g/mol
Boiling Pt:190 °C (6 mmHg)
Melting Pt:131…134 °C
:M-Clarity™ quality level = MQ100
EINECS:225-977-2
Frequently Bought Together