4-Aminoindole
Catalog # MOLE25980595-100MG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
Melting Pt:108 °C
:132.16 g/mol
CAS Number:5192-23-4
Formula:C₈H₈N₂
MDL Number:MFCD01076559