Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
5-Fluoroindole

5-Fluoroindole

Glass bottle for solids, 1 g
Catalog # MOLEM51747925
Irritant
Marketsource
Supplier:   Molekula
5-Fluoroindole
Catalog # MOLEM51747925
Supplier:  Molekula
Marketsource
CAS Number:  399-52-0
5-Fluoroindole

Glass bottle for solids, 1 g

CAS Number:  399-52-0
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  5-Fluoro-1H-indole
  • CAS Number:399-52-0
  • Formula:C₈H₆FN
  • MDL Number:MFCD00005671
  • Molecular Weight:135.14 g/mol
  • Boiling Pt:85 °C (0,5 mmHg)
  • Boiling Pt:85 °C (0.5 mmHg)
  • Melting Pt:46 °C
  • Storage Temperature:Refrigerator
  • EINECS:206-917-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications