2-Ethylimidazole
Glass bottle for solids, 1 kg
Catalog # MOLE23213483-1KG
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2-Ethyl-1H-imidazole
- CAS Number:1072-62-4
- Formula:C₅H₈N₂
- MDL Number:MFCD00005192
- Molecular Weight:96.13 g/mol
- Boiling Pt:268 °C (1013 hPa)
- Density:1,03 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:214-011-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



