Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium sulphate
Ammonium sulphate
Ammonium sulphate
A3598.1000
:
PanReac AppliChem
:
Ammonium sulphate
A3598.1000
:
PanReac AppliChem
:
Pack type:
Glass bottle for solids
Pk:
1 kg
Seq:
4024
:
:
132.14 g/mol
:
13,00559
:
336…339 °C
:
1.78 g/cm³ (20 °C)
:
Ambient
:
231-984-1
:
01-2119455044-46
:
7783-20-2
:
(NH₄)₂SO₄
:
MFCD00003391