Tetraphenylphosphonium chloride
Catalog # FLUO018998-5G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:374.85 g/mol
Melting Pt:275…278 °C
EINECS:217-890-3
CAS Number:2001-45-8
Formula:C₂₄H₂₀ClP
MDL Number:MFCD00011916