Nitronium tetrafluoroborate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13826-86-3
:
Formula:BF₄NO₂
MDL Number:MFCD00011432
:132.81 g/mol
Melting Pt:260 °C
Density:1.26 g/cm³ (20 °C)
EINECS:237-533-5
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ200
Frequently Bought Together