Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Chromic acid 2%

1 L
Catalog # DIUKCH2093
Toxic
Corrosive
Health
Environment
Supplier:   DISTRIBUTOR
Chromic acid 2%
Catalog # DIUKCH2093
Supplier:  DISTRIBUTOR
CAS Number:  7738-94-5
Chromic acid 2%

1 L

CAS Number:  7738-94-5
Price
£164.00
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:7738-94-5
  • Formula:CrH₂O₄
  • Molecular Weight:118.01 g/mol
  • ADR:8,II
  •  :Density: 2.7 g/cm³ (20 °C)
  • UN:1755
Specifications

Specifications

  • Pk:
    1 L

Specifications