

Antimony(III) iodide, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7790-44-5
:
Formula:I₃Sb
MDL Number:MFCD00016322
:502.47 g/mol
Merck Index:14,710
Boiling Pt:420 °C
Melting Pt:168 °C
Density:4.676 g/cm³ (20 °C)
UN:3077
ADR:9,III
:ampouled under argon
EINECS:232-205-8
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together