Phosphorus(III) chloride, Sigma-Aldrich®
Catalog # SIAL310115-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:137.33 g/mol
Merck Index:12,07515
Boiling Pt:76 °C (1013 hPa)
Melting Pt:–112 °C
Density:1.57 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-749-3
UN:1809
ADR:6.1,I
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
:GHS08
CAS Number:7719-12-2
Formula:Cl₃P
MDL Number:MFCD00011438