Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Phenyltriethoxysilane 98%

Phenyltriethoxysilane 98%

Irritant
Flammable
Phenyltriethoxysilane 98%
Certificates
Phenyltriethoxysilane 98%

2 Options Available Below

Product Details & Documents
Synonyms:  Phenyltriethoxysilane, Triethoxy(phenyl)silane, Triethoxyphenylsilane
  • CAS Number:780-69-8
  • Formula:C₁₂H₂₀O₃Si
  • MDL Number:MFCD00009065
  • Molecular Weight:240.37 g/mol
  • Boiling Pt:237 °C (1013 hPa)
  • Melting Pt:41 °C
  • Density:0.996 g/cm³
  • Flash Pt:42 °C
  • EINECS:212-305-8
  • UN:1993

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4594 to 1.4614 (20°C, 589 nm)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - £41.10
    £41.10
    1 * 250 g
       £41.10

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - £126.00
    £126.00
    1 * 1 kg
       £126.00