Order Entry
United Kingdom
ContactUsLinkComponent
Tributyltin chloride
Toxic
Health
Environment
Tributyltin chloride
Catalog # EHERC17150000
Supplier:  EHRENSTORFER
CAS Number:  
Toxic
Health
Environment
Tributyltin chloride
Catalog # EHERC17150000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :325.51 g/mol
Boiling Pt:171…173 °C (25 mmHg)
Melting Pt:–9 °C
Density:1.2 g/cm³ (20 °C)
Flash Pt:>100 °C
EINECS:215-958-7
UN:2788
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:1461-22-9
Formula:C₁₂H₂₇ClSn
MDL Number:MFCD00000521