Dimethyldichlorosilane
Catalog # ABCRAB153237.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
:129.06 g/mol
Boiling Pt:70 °C (1013 hPa)
Melting Pt:–76 °C
Density:1.0745 g/cm³ (20 °C)
Flash Pt:–16 °C
EINECS:200-901-0
UN:1162
ADR:3,II
REACH:01-2119437250-51
:GHS02
:GHS07
CAS Number:75-78-5
Formula:C₂H₆Cl₂Si
MDL Number:MFCD00000491