Tin(IV) iodide, Sigma-Aldrich®
Catalog # SIAL409294-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: Tin tetraiodide
:
:626.33 g/mol
EINECS:232-208-4
UN:3260
CAS Number:7790-47-8
Formula:I₄Sn
MDL Number:MFCD00036272
Melting Pt:143
Density:4.46 g/cm³ (20 °C)
ADR:8 III
Merck Index:13,08855
:M-Clarity™ quality level = MQ100