Barium sulphate 98%, extra pure
:7727-43-7
:
:BaSO₄
:MFCD00003455
:1600
:1580 °C (dec.)
:4.5 g/cm³
:233.39 g/mol
:13,00997
:Ambient
:231-784-4
- Environmentally Preferable:
- Appearance:White Powder
- Assay:97.5 to 100.5 %
- Acidity/alkalinity:Passes test
- Sulfide (S):≤0.5 ppm
- Heavy metals (as Pb):≤10 ppm
- Soluble barium salts:≤10 ppm
- Loss on ignition:≤2.0 % (600°C)
- Oxidisable sulfur compounds:Passes test
- Acid soluble substances:≤0.3 %
Frequently Bought Together