Magnesium iodide, ultra dry ≥99.996% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10377-58-9
:
Formula:MgI₂
MDL Number:MFCD00049485
Density:3.89 g/cm³ (20 °C)
Molecular Weight:278.11 g/mol
Melting Pt:637 °C (dec.)
Merck Index:14,5671
:Ampuled under argon
Storage Temperature:Ambient
EINECS:233-825-1
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together