Order Entry
United Kingdom
ContactUsLinkComponent
Barium perchlorate, Sigma-Aldrich®
Oxidizing
Irritant
Barium perchlorate, Sigma-Aldrich®
Catalog # SIAL241733-100G
Supplier:  Merck
CAS Number:  
Oxidizing
Irritant
Barium perchlorate, Sigma-Aldrich®
Catalog # SIAL241733-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :336.23 g/mol
Merck Index:12,01016
Melting Pt:505 °C
Density:3.2 g/cm³ (20 °C)
Flash Pt:21 °C (closed cup)
EINECS:236-710-4
UN:1447
ADR:5.1,II
 :M-Clarity™ quality level = MQ100
 :GHS03
 :GHS07
CAS Number:13465-95-7
Formula:Ba(ClO₄)₂
MDL Number:MFCD00003440