


Potassium peroxodisulphate
Catalog # HACH245199
Supplier: Hach
CAS Number:
Specifications
- Pk:100
- Seq:0100
Specifications
About this item
:
:270.32 g/mol
Merck Index:13,07741
Melting Pt:100 °C
Density:2.477 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-781-8
UN:1492
ADR:5.1,III
:GHS03
:GHS08
:GHS07
CAS Number:7727-21-1
Formula:K₂S₂O₈
MDL Number:MFCD00011386