Sodium iodide ≥99.5% ACS, Fluka™
:7681-82-5
:
:NaI
:MFCD00003532
:149.89 g/mol
:13,08705
:1.304 °C (1.013 hPa)
:662 °C
:3.67 g/cm³ (20 °C)
:Ambient
:231-679-3
:3077
:9,III
:GHS07
:GHS08
- Assay:Min. 99.5 %
- pH (5 %, 25 °C):6.0 - 9.0
- Water insoluble matter:Max. 0.01 %
- Barium (Ba):Max. 0.002 %
- Calcium (Ca):Max. 0.002 %
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 0.01 %
- Magnesium (Mg):Max. 0.001 %
- Heavy metals (as Pb):Max. 5 ppm
- Chloride and bromide (as Cl):Max. 0.01 %
- Iodate (IO₃):Max. 3 ppm
- Phosphate (PO₄):Max. 0.001 %
- Sulfate (SO₄):Max. 0.005 %
Frequently Bought Together