Sodium iodide ≥99%, Reagent Grade, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7681-82-5
:
Formula:NaI
MDL Number:MFCD00003532
:149.89 g/mol
Merck Index:13,08705
Boiling Pt:1.304 °C (1.013 hPa)
Melting Pt:662 °C
Density:3.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-679-3
ADR:9,III
:GHS07
:GHS08
Specification Test Results
- Assay of I:83.8 - 85.4 %
- Water (Karl Fischer):Max. 1.0 %
Specification Test Results
Frequently Bought Together