Order Entry
United Kingdom
ContactUsLinkComponent
 
Irritant
Cesium carbonate ≥99% (Cs basis)
Catalog # ABCRAB119865.0100
Supplier:  abcr
CAS Number:  
Irritant
Cesium carbonate ≥99% (Cs basis)
Catalog # ABCRAB119865.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :325.82 g/mol
Merck Index:13,02020
Melting Pt:610 °C (dec.)
Density:4.24 g/cm³ (20 °C)
EINECS:208-591-9
 :GHS07
Storage Temperature:Ambient
CAS Number:534-17-8
Formula:Cs₂CO₃
MDL Number:MFCD00010957