Sodium borohydride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16940-66-2
:
Formula:NaBH₄
MDL Number:MFCD00003518
:37.83 g/mol
Merck Index:13,08664
Melting Pt:400 °C
Density:1.07 g/cm³ (20 °C)
Flash Pt:69 °C
Storage Temperature:Ambient
UN:1426
ADR:4.3,I
:M-Clarity™ quality level = MQ200
EINECS:241-004-4
:GHS02
:GHS06
:GHS05
Frequently Bought Together