Lithium tetrahydroborate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16949-15-8
:
Formula:H₄BLi
MDL Number:MFCD00011088
:21.78 g/mol
Merck Index:13,05547
Boiling Pt:66 °C (1013 hPa)
Melting Pt:274…277 °C
Density:0.666 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1413
ADR:4.3,I
:M-Clarity™ quality level = MQ100
EINECS:241-021-7
Frequently Bought Together