

Sodium hexafluoroarsenate ≥99%
Catalog # 88517.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
CAS Number:12005-86-6
:
Formula:AsF₆Na
MDL Number:MFCD00014228
:211.9 g/mol
UN:1557
ADR:6.1,II
:GHS06
:GHS09
Storage Temperature:Ambient