Sodium hexafluoroaluminate, Sigma-Aldrich®
SIAL305499-500G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
: Trisodium hexafluoroaluminate, Sodium hexafluoroaluminate(III)
:
:209.94 g/mol
:977 °C
:2.95 g/cm³ (20 °C)
:8,III
:M-Clarity™ quality level = MQ100
:237-410-6
:3260
:GHS08
:GHS07
:GHS09
:13775-53-6
:15096-52-3
:AlF₆Na₃
:MFCD00003507