2,7-Dinitro-9-fluorenone
Glass bottle for solids, 25 g
Catalog # MOLEM12944437
Marketsource
Supplier: Molekula
2,7-Dinitro-9-fluorenone
Catalog # MOLEM12944437
Supplier: Molekula
Marketsource
CAS Number: 31551-45-8
Jump to:
Product Details & Documents
- CAS Number:31551-45-8
- Formula:C₁₃H₆N₂O₅
- MDL Number:MFCD00001153
- Molecular Weight:270.2 g/mol
- Melting Pt:296 °C
- EINECS:250-695-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



