Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
9-Fluorenone

9-Fluorenone

Glass bottle for solids, 500 g
Catalog # MOLE30859177-500G
Marketsource
Supplier:   Molekula
9-Fluorenone
Catalog # MOLE30859177-500G
Supplier:  Molekula
Marketsource
CAS Number:  486-25-9
9-Fluorenone

Glass bottle for solids, 500 g

CAS Number:  486-25-9
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  9H-Fluoren-9-one
  • CAS Number:486-25-9
  • Formula:C₁₃H₈O
  • MDL Number:MFCD00001141
  • Molecular Weight:180.21 g/mol
  • Boiling Pt:342 °C (1013 hPa)
  • Melting Pt:81…83 °C
  • Density:0.9 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-630-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.