9-Fluorenone 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 9H-Fluoren-9-one
- CAS Number:486-25-9
- Formula:C₁₃H₈O
- MDL Number:MFCD00001141
- Molecular Weight:180.21 g/mol
- Boiling Pt:342 °C (760 mmHg)
- Melting Pt:81…85 °C
- Density:0,9 g/cm³ (20 °C)
- Flash Pt:163 °C
- Storage Temperature:Ambient
- EINECS:207-630-7
- UN:3077
3 Options Available Below
Product Details & Documents
Synonyms: 9H-Fluoren-9-one
- CAS Number:486-25-9
- Formula:C₁₃H₈O
- MDL Number:MFCD00001141
- Molecular Weight:180.21 g/mol
- Boiling Pt:342 °C (760 mmHg)
- Melting Pt:81…85 °C
- Density:0,9 g/cm³ (20 °C)
- Flash Pt:163 °C
- Storage Temperature:Ambient
- EINECS:207-630-7
- UN:3077
Documents
ACRO11920
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Yellow Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:81 to 85 °C
- GC:≥99.0 %
- NMR (1H-NMR):Consistent with structure
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



