Order Entry
United Kingdom
ContactUsLinkComponent
 
 
 
Octamethylcyclotetrasiloxane 98%
 
undefined
 
 
Octamethylcyclotetrasiloxane 98%
 

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3950 to 1.3970 (20°C, 589 nm)
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together