Cyclopropanecarbonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4023-34-1
:
Formula:C₄H₅ClO
MDL Number:MFCD00001277
:104.54 g/mol
Boiling Pt:119 °C (1013 hPa)
Density:1.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:223-684-4
UN:2920
ADR:8,II
:M-Clarity™ quality level = MQ200
Frequently Bought Together